Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichlorothiophene 1,1-Dioxide 96.0+%, TCI America™
SDP

Catalog No. D36121G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D36121G Supplier TCI America Supplier No. D36121G

Chemical Identifiers

CAS 52819-14-4
Molecular Formula C4H2Cl2O2S
Molecular Weight (g/mol) 185.018
InChI Key CIDAIMONSYOOGS-UHFFFAOYSA-N
PubChem CID 3040679
IUPAC Name 3,4-dichlorothiophene 1,1-dioxide
SMILES C1=C(C(=CS1(=O)=O)Cl)Cl

Specifications

Melting Point 108°C
Color White-Yellow
Quantity 1 g
Formula Weight 185.02
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dichlorothiophene 1,1-Dioxide
RTECSNumber XM8650000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.