CAS RN 52831-11-5
IUPAC Name:
1,3,5-trimethyl cyclohexane-1,3,5-tricarboxylate
Synonyms:
1,3,5-trimethyl cyclohexane-1,3,5-tricarboxylate
Molecular Weight (g/mol):
258.27
Molecular Formula:
C12H18O6
InChi Key:
ZZNOMLSSCRBRJS-UHFFFAOYSA-N
SMILES:
COC(=O)C1CC(CC(C1)C(=O)OC)C(=O)OC
1
–
1
of
1
results
1
Trimethyl 1,3,5-Cyclohexanetricarboxylate (cis- and trans- mixture) 98.0+%, TCI America™
CAS: 52831-11-5 Molecular Formula: C12H18O6 Molecular Weight (g/mol): 258.27 InChI Key: ZZNOMLSSCRBRJS-UHFFFAOYSA-N Synonym: 1,3,5-Cyclohexanetricarboxylic Acid Trimethyl Ester PubChem CID: 247401 IUPAC Name: trimethyl cyclohexane-1,3,5-tricarboxylate SMILES: COC(=O)C1CC(CC(C1)C(=O)OC)C(=O)OC