accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3CH3

CAS RN 52890-52-5

IUPAC Name: phenyl(2,4,5-trimethylphenyl)methanone
Synonyms: phenyl(2,4,5-trimethylphenyl)methanone
Molecular Weight (g/mol): 224.3
Molecular Formula: C16H16O
InChi Key: FTPZYWUJDVZEIE-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C)C=C1C(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results