Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trimethylbenzophenone 95.0+%, TCI America™
SDP

Catalog No. T20621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T20621G Supplier TCI America Supplier No. T20621G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52890-52-5
Molecular Formula C16H16O
Molecular Weight (g/mol) 224.303
InChI Key FTPZYWUJDVZEIE-UHFFFAOYSA-N
PubChem CID 14907151
IUPAC Name phenyl-(2,4,5-trimethylphenyl)methanone
SMILES CC1=CC(=C(C=C1C(=O)C2=CC=CC=C2)C)C

Specifications

Color Yellow
Boiling Point 328°C
Quantity 1 g
Formula Weight 224.30
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,5-Trimethylbenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.