accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CNOO

CAS RN 52898-32-5

IUPAC Name: 2-(but-3-en-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(but-3-en-1-yl)isoindole-1,3-dione
Molecular Weight (g/mol): 201.23
Molecular Formula: C12H11NO2
InChi Key: ZDOLXCKKXHSEJG-UHFFFAOYSA-N
SMILES: C=CCCN1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results