Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(3-Buten-1-yl)phthalimide 98.0+%, TCI America™
SDP

Catalog No. B18815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B18815G Supplier TCI America Supplier No. B18815G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52898-32-5
Molecular Formula C12H11NO2
Molecular Weight (g/mol) 201.225
MDL Number MFCD00175471
InChI Key ZDOLXCKKXHSEJG-UHFFFAOYSA-N
PubChem CID 339432
IUPAC Name 2-but-3-enylisoindole-1,3-dione
SMILES C=CCCN1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 52°C
Color White
Quantity 5 g
Formula Weight 201.23
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(3-Buten-1-yl)phthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.