accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH

CAS RN 529-32-8

IUPAC Name: decahydronaphthalen-1-ol
Synonyms: decahydronaphthalen-1-ol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: NDZOISQLWLWLEW-UHFFFAOYNA-N
SMILES: OC1CCCC2CCCCC12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results