accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 529-34-0

IUPAC Name: 1,2,3,4-tetrahydronaphthalen-1-one
Synonyms: 1-tetralone
Molecular Weight (g/mol): 146.19
Molecular Formula: C10H10O
InChi Key: XHLHPRDBBAGVEG-UHFFFAOYSA-N
SMILES: O=C1CCCC2=CC=CC=C12
Boiling Point 
  • (2)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (4)
  • (3)
Quantity 
  • (3)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (4)
  • (5)