Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Tetralone, 98%

Catalog No. AC138310250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC138310250 25 g Glass bottle
AC138311000 100 g Glass bottle
AC138315000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC138310250 Supplier Thermo Scientific Chemicals Supplier No. 138310250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 529-34-0 | C10H10O | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 529-34-0
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00001688
InChI Key XHLHPRDBBAGVEG-UHFFFAOYSA-N
Synonym alpha-tetralone, 1-tetralone, 3,4-dihydronaphthalen-1 2h-one, 1,2,3,4-tetrahydronaphthalen-1-one, 3,4-dihydro-1 2h-naphthalenone, tetralone, a-tetralone, 1-oxotetralin, 1 2h-naphthalenone, 3,4-dihydro, .alpha.-tetralone
PubChem CID 10724
IUPAC Name 3,4-dihydro-2H-naphthalen-1-one
SMILES O=C1CCCC2=CC=CC=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 5.0°C to 6.0°C
Color Brown to Yellow
Density 1.0960g/mL
Boiling Point 127.0°C (13.0 mmHg)
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5675 to 1.5695
Quantity 25 g
Beilstein 07, 370
Specific Gravity 1.096
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hexane
Formula Weight 146.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material α-Tetralone, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-460-6
RTECSNumber QK4375000
TSCA TSCA
Recommended Storage Tends to darken during storage

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.