accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CHNONHH2N

CAS RN 5294-61-1

IUPAC Name: 4-{[(2,6-dimethylphenyl)carbamoyl]methyl}piperazine-1,4-diium
Synonyms: 4-{[(2,6-dimethylphenyl)carbamoyl]methyl}piperazine-1,4-diium
Molecular Weight (g/mol): 249.36
Molecular Formula: C14H23N3O
InChi Key: NJKRFQIWDJSYOK-UHFFFAOYSA-P
SMILES: CC1=CC=CC(C)=C1NC(=O)C[NH+]1CC[NH2+]CC1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)