Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2,6-Dimethylphenyl)-1-piperazineacetamide 98.0+%, TCI America™
SDP

Catalog No. D431925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D431925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D431925G Supplier TCI America Supplier No. D431925G

missing translation for 'validMainframeMsg'

Specifications

CAS 5294-61-1
Melting Point 118°C
Color White
Molecular Formula C14H21N3O
Quantity 25 g
MDL Number MFCD06384973
Synonym n-2,6-dimethylphenyl-2-piperazin-1-yl acetamide, n-2,6-diphenylmethyl-1-piperazine acetylamine, n-2,6-dimethylphenyl-1-piperazineacetamide, n-2,6-dimethylphenyl-2-piperazin-1-ylacetamide, unii-nys3i6283h, 1-piperazineacetamide, n-2,6-dimethylphenyl, 1-n-2,6-dimethylphenyl carbamoylmethyl piperazine, n-2,6-dimethylphenyl-2-piperazinylacetamide, n-2,6-dimethylphenyl-2-piperazin-1-yl-acetamide, 4-2,6-dimethylphenyl aminocarbonylmethyl piperazine
InChI Key NJKRFQIWDJSYOK-UHFFFAOYSA-N
SMILES CC1=C(C(=CC=C1)C)NC(=O)CN2CCNCC2
IUPAC Name N-(2,6-dimethylphenyl)-2-piperazin-1-ylacetamide
Molecular Weight (g/mol) 247.342
PubChem CID 2608606
Formula Weight 247.34
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2,6-Dimethylphenyl)-1-piperazineacetamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.