accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52944-34-0

CH3Cl

CAS RN 52944-34-0

IUPAC Name: 1-chloro-4-propylbenzene
Synonyms: 1-chloro-4-propylbenzene
Molecular Weight (g/mol): 154.64
Molecular Formula: C9H11Cl
InChi Key: QXQAPNSHUJORMC-UHFFFAOYSA-N
SMILES: CCCC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results