Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-4-propylbenzene 98.0+%, TCI America™
SDP

Catalog No. C20875G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20875G Supplier TCI America Supplier No. C20875G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52944-34-0
Molecular Formula C9H11Cl
Molecular Weight (g/mol) 154.64
MDL Number MFCD00270099
InChI Key QXQAPNSHUJORMC-UHFFFAOYSA-N
Synonym 4-Propylchlorobenzene
PubChem CID 521435
IUPAC Name 1-chloro-4-propylbenzene
SMILES CCCC1=CC=C(Cl)C=C1

Specifications

Color Yellow
Boiling Point 203°C
Quantity 5 g
Formula Weight 154.64
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-4-propylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.