accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52950-19-3

OOHHOCl

CAS RN 52950-19-3

IUPAC Name: 2-(2-chlorophenyl)-2-hydroxyacetic acid
Synonyms: (2-chlorophenyl)(hydroxy)acetic acid
Molecular Weight (g/mol): 186.59
Molecular Formula: C8H7ClO3
InChi Key: RWOLDZZTBNYTMS-UHFFFAOYNA-N
SMILES: OC(C(O)=O)C1=CC=CC=C1Cl
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)