Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-Chloromandelic acid, 95%

Catalog No. AAH6115214
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6115214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6115214 Supplier Thermo Scientific Chemicals Supplier No. H6115214

missing translation for 'validMainframeMsg'

CAS: 52950-19-3 | C8H7ClO3 | 186.591 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52950-19-3
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00798437
InChI Key RWOLDZZTBNYTMS-ZETCQYMHSA-N
Synonym s-+-2-chloromandelic acid, s-2-2-chlorophenyl-2-hydroxyacetic acid, s-2-chloromandelic acid, s-o-chloromandelicacid, 2-chloro-l-mandelic acid, 2s-2-2-chlorophenyl-2-hydroxy-acetic acid, s-2-chlorophenyl hydroxy acetic acid, pubchem5743, s-o-chloromandelic acid, ksc914s2j
PubChem CID 6922874
IUPAC Name (2S)-2-(2-chlorophenyl)-2-hydroxyacetic acid
SMILES C1=CC=C(C(=C1)C(C(=O)O)O)Cl

Specifications

Melting Point 118°C to 122°C
Color Brown to White
Quantity 25 g
Physical Form Powder
Formula Weight 186.59
Percent Purity 95%
Chemical Name or Material (S)-(+)-2-Chloromandelic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.