CAS RN 52988-34-8
IUPAC Name:
4'-methoxy-[1,1'-biphenyl]-4-carbaldehyde
Synonyms:
4'-methoxy-[1,1'-biphenyl]-4-carbaldehyde
Molecular Weight (g/mol):
212.25
Molecular Formula:
C14H12O2
InChi Key:
JTTIGLYPLMYHAT-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)C1=CC=C(C=O)C=C1
1
–
1
of
1
results
1
Sigma Aldrich 4-(4-methoxyphenyl)benzaldehyde
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.