accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5299-60-5

IUPAC Name: ethyl 6-hydroxyhexanoate
Synonyms: ethyl 6-hydroxyhexanoate
Molecular Weight (g/mol): 160.21
Molecular Formula: C8H16O3
InChi Key: HYXRUZUPCFVWAH-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCCO
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results