accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3COCH3(E)

CAS RN 52993-95-0

IUPAC Name: sodium 1-methoxy-4-[(1E)-prop-1-en-1-yl]benzene
Synonyms: sodium anethole
Molecular Weight (g/mol): 171.19
Molecular Formula: C10H12NaO
InChi Key: CZGYTOCDSMDCRG-BJILWQEISA-N
SMILES: [Na+].COC1=CC=C(\C=C\C)C=C1
Grade 
  • (4)
  • (4)
Physical Form 
  • (4)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)