accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)(S)(S)(R)OHOOCH3O(S)CH3(S)HO(S)(R)CH3O

CAS RN 53-36-1

IUPAC Name: 2-[(1R,3aS,3bS,5S,9aR,9bS,10S,11aS)-1,10-dihydroxy-5,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
Synonyms: depo-medrol demethyl methylprednisolone acetate D-med 21-acetate depo-medrone methylprednisolone-21-acetate methylprednisolone 21-acetate mepred M-predrol
Molecular Weight (g/mol): 416.51
Molecular Formula: C24H32O6
InChi Key: PLBHSZGDDKCEHR-LFYFAGGJSA-N
SMILES: C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)COC(C)=O)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]2(C)C=CC(=O)C=C12
Grade 
  • (1)
  • (6)
Percent Purity 
  • (6)
Physical Form 
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (1)
  • (6)
  • (6)
  • (1)
  • (6)
  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)