Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylprednisolone Acetate, Micronized, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18604543 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
250 g
1 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-543 250 g Amber Glass Bottle
18-604-538 1 g Amber Glass Bottle
18-604-539 5 g Amber Glass Bottle
18-604-540 25 g Amber Glass Bottle
18-604-542 100 g Amber Glass Bottle
18-604-541 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-604-543 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ME171250GMBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ME171, 53-36-1, C24H32O6

Spectrum™ Chemical Methylprednisolone Acetate, Micronized, USP is a synthetic glucocorticoid typically used as an anti-inflammatory. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 53-36-1
Molecular Formula C24H32O6
Molecular Weight (g/mol) 416.51
InChI Key PLBHSZGDDKCEHR-LFYFAGGJSA-N
IUPAC Name 2-[(1R,3aS,3bS,5S,9aR,9bS,10S,11aS)-1,10-dihydroxy-5,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
SMILES C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)COC(C)=O)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]2(C)C=CC(=O)C=C12

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 250 g
Specific Rotation +97° to +105°
Percent Purity 97 to 103%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Methylprednisolone Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.