CAS RN 53-84-9
beta-Nicotinamide adenine dinucleotide hydrate, 98+%
CAS: 53-84-9 Molecular Formula: C21H27N7O14P2 Molecular Weight (g/mol): 663.43 MDL Number: MFCD00150377 InChI Key: BAWFJGJZGIEFAR-WIWLTUSXNA-N Synonym: nicotinamide adenine dinucleotide,nad+,diphosphopyridine nucleotide,nad-oxidized,nicotinamide-adenine dinucleotide,dpn-ox,beta-nicotinamide adenine dinucleotide,dpn+,nad,nad + PubChem CID: 15938971 ChEBI: CHEBI:57540 IUPAC Name: [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate SMILES: NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
beta-Nicotinamide adenine dinucleotide, 97%
CAS: 53-84-9 Molecular Formula: C21H27N7O14P2 Molecular Weight (g/mol): 663.43 MDL Number: MFCD00150377 InChI Key: BAWFJGJZGIEFAR-WIWLTUSXNA-N Synonym: nicotinamide adenine dinucleotide,nad+,diphosphopyridine nucleotide,nad-oxidized,nicotinamide-adenine dinucleotide,dpn-ox,beta-nicotinamide adenine dinucleotide,dpn+,nad,nad + PubChem CID: 15938971 ChEBI: CHEBI:57540 IUPAC Name: [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate SMILES: NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
β-Nicotinamide Adenine Dinucleotide, MP Biomedicals™
CAS: 53-84-9 Molecular Formula: C21H27N7O14P2 Molecular Weight (g/mol): 663.43 MDL Number: MFCD00150377 InChI Key: BAWFJGJZGIEFAR-WIWLTUSXNA-N Synonym: nicotinamide adenine dinucleotide,nad+,diphosphopyridine nucleotide,nad-oxidized,nicotinamide-adenine dinucleotide,dpn-ox,beta-nicotinamide adenine dinucleotide,dpn+,nad,nad + PubChem CID: 15938971 ChEBI: CHEBI:57540 IUPAC Name: [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate SMILES: NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
beta-Nicotinamide Adenine Dinucleotide Hydrate, oxidized form 95.0+%, TCI America™
CAS: 53-84-9 Molecular Formula: C21H27N7O14P2 Molecular Weight (g/mol): 663.43 MDL Number: MFCD00150377 InChI Key: BAWFJGJZGIEFAR-WIWLTUSXNA-N Synonym: nicotinamide adenine dinucleotide,nad+,diphosphopyridine nucleotide,nad-oxidized,nicotinamide-adenine dinucleotide,dpn-ox,beta-nicotinamide adenine dinucleotide,dpn+,nad,nad + PubChem CID: 15938971 ChEBI: CHEBI:57540 IUPAC Name: 1-[(2R,3R,4S,5R)-5-{[({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]methyl}-3,4-dihydroxyoxolan-2-yl]-3-carbamoyl-1λ⁵-pyridin-1-ylium SMILES: NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
β-Nicotinamide Adenine Dinucleotide, Oxidized, Free Acid, 99%, MP Biomedicals™
CAS: 53-84-9 Molecular Formula: C21H27N7O14P2 Molecular Weight (g/mol): 663.43 MDL Number: MFCD00150377 InChI Key: BAWFJGJZGIEFAR-WIWLTUSXNA-N Synonym: nicotinamide adenine dinucleotide,nad+,diphosphopyridine nucleotide,nad-oxidized,nicotinamide-adenine dinucleotide,dpn-ox,beta-nicotinamide adenine dinucleotide,dpn+,nad,nad + PubChem CID: 15938971 ChEBI: CHEBI:57540 IUPAC Name: [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate SMILES: NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
b-Nicotinamide Adenine Dinucleotide, 95%, Spectrum™ Chemical
CAS: 53-84-9 Molecular Formula: C21H27N7O14P2 Molecular Weight (g/mol): 663.43 MDL Number: MFCD00150377 InChI Key: BAWFJGJZGIEFAR-WIWLTUSXNA-N IUPAC Name: 1-[(2R,3R,4S,5R)-5-{[({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]methyl}-3,4-dihydroxyoxolan-2-yl]-3-carbamoyl-1λ5-pyridin-1-ylium SMILES: NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
β-Nicotinamide Adenine Dinucleotide, MP Biomedicals™
CAS: 53-84-9 Molecular Formula: C21H27N7O14P2 Molecular Weight (g/mol): 663.43 MDL Number: MFCD00150377 InChI Key: BAWFJGJZGIEFAR-WIWLTUSXNA-N Synonym: nicotinamide adenine dinucleotide,nad+,diphosphopyridine nucleotide,nad-oxidized,nicotinamide-adenine dinucleotide,dpn-ox,beta-nicotinamide adenine dinucleotide,dpn+,nad,nad + PubChem CID: 15938971 ChEBI: CHEBI:57540 IUPAC Name: [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate SMILES: NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
β-Nicotinamide adenine dinucleotide phosphate, Cell Culture Reagent, MP Biomedicals™
CAS: 53-84-9 Molecular Formula: C21H27N7O14P2 Molecular Weight (g/mol): 663.43 MDL Number: MFCD00150377 InChI Key: BAWFJGJZGIEFAR-WIWLTUSXNA-N Synonym: nicotinamide adenine dinucleotide,nad+,diphosphopyridine nucleotide,nad-oxidized,nicotinamide-adenine dinucleotide,dpn-ox,beta-nicotinamide adenine dinucleotide,dpn+,nad,nad + PubChem CID: 15938971 ChEBI: CHEBI:57540 SMILES: NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
β-Nicotinamide adenine dinucleotide, oxidized Form, free acid, >98%, MP Biomedicals™
CAS: 53-84-9 Molecular Formula: C21H27N7O14P2 Molecular Weight (g/mol): 663.43 MDL Number: MFCD00150377 InChI Key: BAWFJGJZGIEFAR-WIWLTUSXNA-N Synonym: nicotinamide adenine dinucleotide,nad+,diphosphopyridine nucleotide,nad-oxidized,nicotinamide-adenine dinucleotide,dpn-ox,beta-nicotinamide adenine dinucleotide,dpn+,nad,nad + PubChem CID: 15938971 ChEBI: CHEBI:57540 IUPAC Name: [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate SMILES: NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O