accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53-96-3

IUPAC Name: N-(9H-fluoren-2-yl)acetamide
Synonyms: 2 acetylaminofluorene 2-aaf 2-faa
Molecular Weight (g/mol): 223.28
Molecular Formula: C15H13NO
InChi Key: CZIHNRWJTSTCEX-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C2C(CC3=CC=CC=C23)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results