Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetamidofluorene 98.0+%, TCI America™
SDP

Catalog No. A00765G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A00765G 5 g
A007625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A00765G Supplier TCI America Supplier No. A00765G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53-96-3
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.28
MDL Number MFCD00001116
InChI Key CZIHNRWJTSTCEX-UHFFFAOYSA-N
Synonym 2-acetamidofluorene, 2-acetylaminofluorene, n-2-fluorenylacetamide, 2-aaf, n-9h-fluoren-2-yl acetamide, acetamide, n-9h-fluoren-2-yl, n-fluoren-2-ylacetamide, n-acetyl-2-aminofluorene, 2-acetaminofluorene, 2-acetylamino fluorene
PubChem CID 5897
ChEBI CHEBI:17356
IUPAC Name N-(9H-fluoren-2-yl)acetamide
SMILES CC(=O)NC1=CC=C2C(CC3=CC=CC=C23)=C1

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 5 g
Formula Weight 223.28
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamidofluorene
RTECSNumber AB9450000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.