CAS RN 530-26-7
IUPAC Name:
(3S,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Synonyms:
β-glucose blood sugar β-dextrose β-mannose D glucose hexose (L)-glucose (l)-glucose D mannose L glucose
Molecular Weight (g/mol):
180.16
Molecular Formula:
C6H12O6
InChi Key:
WQZGKKKJIJFFOK-QTVWNMPRSA-N
SMILES:
OC[C@H]1OC(O)[C@@H](O)[C@@H](O)[C@@H]1O
1
–
1
of
1
results
1
MP Biomedicals, Inc D-Mannose, [2-3H] , MP Biomedical
CAS: 530-26-7 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.16 InChI Key: WQZGKKKJIJFFOK-QTVWNMPRSA-N Synonym: d-mannose,mannose,d-mannopyranose,carubinose,seminose,mannopyranose,mannopyranoside,dl-mannose,d +-mannose,+--mannose PubChem CID: 18950 ChEBI: CHEBI:4208