Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Mannose, [2-3H] , MP Biomedical

Catalog No. MP127084S01 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mci
250 μci
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP127084S01 1 mci
MP0127084S2 250 μci
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP127084S01 Supplier MP Biomedicals, Inc Supplier No. 0127084S01

Sterile aqueous solution. HPLC Analyzed for Purity.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-26-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key WQZGKKKJIJFFOK-QTVWNMPRSA-N
Synonym d-mannose, mannose, d-mannopyranose, carubinose, seminose, mannopyranose, mannopyranoside, dl-mannose, d +-mannose, +--mannose
PubChem CID 18950
ChEBI CHEBI:4208

Specifications

Quantity 1 mci
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.