accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 530-43-8

H3COO(R)NHOClCl(R)HONOO

CAS RN 530-43-8

IUPAC Name: (2R,3R)-2-(2,2-dichloroacetamido)-3-hydroxy-3-(4-nitrophenyl)propyl hexadecanoate
Synonyms: chloramphenicol palmitate detreopal chloramphenicol monopalmitate chloromycetin palmitate quemicetina cap-palmitate
Molecular Weight (g/mol): 561.54
Molecular Formula: C27H42Cl2N2O6
InChi Key: PXKHGMGELZGJQE-ILBGXUMGSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OC[C@@H](NC(=O)C(Cl)Cl)[C@H](O)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)