CAS RN 5300-03-8
9-cis-Retinoic acid
CAS: 5300-03-8 Molecular Formula: C20H28O2 Molecular Weight (g/mol): 300.44 MDL Number: MFCD00270072 InChI Key: SHGAZHPCJJPHSC-YCNIQYBTSA-N Synonym: alitretinoin,9-cis-retinoic acid,panretin,9-cis-tretinoin,panrexin,panretyn,panretin gel,toctino,9-cis retinoic acid,9 z-retinoic acid PubChem CID: 449171 ChEBI: CHEBI:50648 IUPAC Name: (2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid SMILES: CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C
9-cis-Retinoic Acid, >98%, MP Biomedicals™
Molecular Formula: C20H28O2 Molecular Weight (g/mol): 300.44 InChI Key: SHGAZHPCJJPHSC-YCNIQYBTSA-N ChEBI: CHEBI:50648