accessibility menu, dialog, popup

UserName

Viewing profile as user:
FBrFFF

CAS RN 53001-71-1

IUPAC Name: 1-(bromomethyl)-2,3,4,5-tetrafluorobenzene
Synonyms: 1-(bromomethyl)-2,3,4,5-tetrafluorobenzene
Molecular Weight (g/mol): 243
Molecular Formula: C7H3BrF4
InChi Key: WTKVWOHBTGWNEO-UHFFFAOYSA-N
SMILES: FC1=CC(CBr)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results