accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53001-73-3

IUPAC Name: 3-(bromomethyl)-1,2,4,5-tetrafluorobenzene
Synonyms: 3-(bromomethyl)-1,2,4,5-tetrafluorobenzene
Molecular Weight (g/mol): 243
Molecular Formula: C7H3BrF4
InChi Key: QHZBSXQCGIQSID-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(F)C(CBr)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results