Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrafluorobenzyl Bromide 97.0+%, TCI America™
SDP

Catalog No. T18865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18865G 5 g
T18861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18865G Supplier TCI America Supplier No. T18865G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53001-73-3
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 242.999
MDL Number MFCD00013549
InChI Key QHZBSXQCGIQSID-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluorobenzyl bromide, 3-bromomethyl-1,2,4,5-tetrafluorobenzene, 2,3,5,6-tetrafluorobenzylbromide, benzene, 3-bromomethyl-1,2,4,5-tetrafluoro, alpha-bromo-2,3,5,6-tetrafluorotoluene, pubchem7361, acmc-1b1z8, benzene,3-bromomethyl-1,2,4,5-tetrafluoro
PubChem CID 104377
IUPAC Name 3-(bromomethyl)-1,2,4,5-tetrafluorobenzene
SMILES C1=C(C(=C(C(=C1F)F)CBr)F)F

Specifications

Color Yellow
Boiling Point 80°C
Quantity 5 g
UN Number 3265
Formula Weight 243.00
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,5,6-Tetrafluorobenzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.