accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHOOOO

CAS RN 53008-65-4

IUPAC Name: [3,4,5-tris(benzyloxy)-6-methoxyoxan-2-yl]methanol
Synonyms: [3,4,5-tris(benzyloxy)-6-methoxyoxan-2-yl]methanol
Molecular Weight (g/mol): 464.56
Molecular Formula: C28H32O6
InChi Key: MOKYEUQDXDKNDX-UHFFFAOYNA-N
SMILES: COC1OC(CO)C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results