accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53066-19-6

IUPAC Name: 1-(2,6-dichlorophenyl)ethan-1-ol
Synonyms: 1-(2,6-dichlorophenyl)ethanol
Molecular Weight (g/mol): 191.05
Molecular Formula: C8H8Cl2O
InChi Key: VUSOJMQVQGKPNN-UHFFFAOYNA-N
SMILES: CC(O)C1=C(Cl)C=CC=C1Cl
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results