Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2,6-Dichlorophenyl)ethanol, 97%, Thermo Scientific™

Catalog No. AAL1235003
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1235003 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1235003 Supplier Thermo Scientific Chemicals Supplier No. L1235003

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 53066-19-6
Boiling Point 117°C to 118°C (4mmHg)
Molecular Formula C8H8Cl2O
Refractive Index 1.567
Quantity 1g
MDL Number MFCD00016853
Beilstein 3243202
Synonym 1-2,6-dichlorophenyl ethanol, 2,6-dichloro-alpha-methylbenzyl alcohol, acmc-209uny, 2,6-dichlorophenylethanol, 1-2,6-dichlorophenyl-ethanol, 2,6-dichlorophenyl methyl carbinol, 1-2,6-dichlorophenyl ethan-1-ol, racemic 1-2,6-dichlorophenyl ethanol, benzenemethanol,2,6-dichloro-a-methyl, benzenemethanol, 2,6-dichloro-.alpha.-methyl
InChI Key VUSOJMQVQGKPNN-UHFFFAOYNA-N
SMILES CC(O)C1=C(Cl)C=CC=C1Cl
IUPAC Name 1-(2,6-dichlorophenyl)ethanol
Molecular Weight (g/mol) 191.05
PubChem CID 103731
Formula Weight 191.06
Percent Purity 97%
Chemical Name or Material 1-(2,6-Dichlorophenyl)ethanol
Show More Show Less
EINECSNumber 258-340-2
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.