accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrNH2

CAS RN 53078-85-6

IUPAC Name: 2-bromo-5-methylaniline
Synonyms: 2-bromo-5-methylaniline
Molecular Weight (g/mol): 186.05
Molecular Formula: C7H8BrN
InChi Key: QTAQWOXSUFGGKH-UHFFFAOYSA-N
SMILES: CC1=CC(N)=C(Br)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)