accessibility menu, dialog, popup

UserName

Viewing profile as user:
OBr

CAS RN 53087-13-1

IUPAC Name: 1-(benzyloxy)-3-bromobenzene
Synonyms: 1-(benzyloxy)-3-bromobenzene
Molecular Weight (g/mol): 263.13
Molecular Formula: C13H11BrO
InChi Key: HVWZMGZBJCJDOX-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(OCC2=CC=CC=C2)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)