Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyloxy-3-bromobenzene 98.0+%, TCI America™
SDP

Catalog No. B23075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23075G 5 g
B230725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23075G Supplier TCI America Supplier No. B23075G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53087-13-1
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.134
MDL Number MFCD00155065
InChI Key HVWZMGZBJCJDOX-UHFFFAOYSA-N
Synonym 1-benzyloxy-3-bromobenzene, 3-benzyloxybromobenzene, benzyl 3-bromophenyl ether, 1-bromo-3-benzyloxybenzene, 3-benzyloxy bromobenzene, 3-bromophenyl benzyl ether, benzene, 1-bromo-3-phenylmethoxy, 1-benzyloxy-3-bromo-benzene, pubchem3065
PubChem CID 2756638
IUPAC Name 1-bromo-3-phenylmethoxybenzene
SMILES C1=CC=C(C=C1)COC2=CC(=CC=C2)Br

Specifications

Melting Point 62°C
Color White
Quantity 5 g
Formula Weight 263.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyloxy-3-bromobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.