accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 532-12-7

IUPAC Name: 3-(3,4-dihydro-2H-pyrrol-5-yl)pyridine
Synonyms: myosmine
Molecular Weight (g/mol): 146.19
Molecular Formula: C9H10N2
InChi Key: DPNGWXJMIILTBS-UHFFFAOYSA-N
SMILES: C1CN=C(C1)C1=CC=CN=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)