accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 532-82-1

IUPAC Name: hydrogen 4-(2-phenyldiazen-1-yl)benzene-1,3-diamine chloride
Synonyms: chrysoidine monohydrochloride
Molecular Weight (g/mol): 248.71
Molecular Formula: C12H13ClN4
InChi Key: MCTQNEBFZMBRSQ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=C(N=NC2=CC=CC=C2)C(N)=C1
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results