Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Chrysoidin, pure

Catalog No. AC200740500
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC200740500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC200740500 Supplier Thermo Scientific Chemicals Supplier No. 200740500
  • $69.75
    $56.65 / Each of 1

    Save $13.10

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 532-82-1 | C12H12N4·HCl | 248.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-82-1
Molecular Formula C12H12N4·HCl
Molecular Weight (g/mol) 248.72
MDL Number MFCD00012976
InChI Key MCTQNEBFZMBRSQ-UHFFFAOYSA-N
Synonym chrysoidine, basic orange 2, chrysoidine g, chrysoidine y, chrysoidin, c.i. basic orange 2, chrysoidine ygh, chrysoidine a, chrysoidine b, chrysoidine j
PubChem CID 10771
IUPAC Name 4-phenyldiazenylbenzene-1,3-diamine;hydrochloride
SMILES C1=CC=C(C=C1)N=NC2=C(C=C(C=C2)N)N.Cl

Specifications

Melting Point 235.0°C
Color Bordeaux to Purple
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 50 g
Beilstein 16, 383
Merck Index 15, 2261
Solubility Information Solubility in water: 55g/L (15°C). Other solubilities: 4.75% absolute ethanol - 6.0% cellosolve, 9.5% anhydr.ethylene glycol, slightly soluble in acetone, practically insoluble in benzene, xylene and ether
Formula Weight 248.72
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material Chrysoidin
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Suspected of causing genetic defects.
Causes serious eye damage.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
EINECSNumber 208-545-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.