CAS RN 532-91-2
IUPAC Name:
6-methoxy-2,3-dihydro-1,3-benzoxazol-2-one
Synonyms:
6-methoxy-3H-1,3-benzoxazol-2-one
Molecular Weight (g/mol):
165.15
Molecular Formula:
C8H7NO3
InChi Key:
MKMCJLMBVKHUMS-UHFFFAOYSA-N
SMILES:
COC1=CC=C2NC(=O)OC2=C1