accessibility menu, dialog, popup

UserName

Viewing profile as user:
ICH3NOONCH3(Z)(E/Z)

CAS RN 53213-82-4

IUPAC Name: 3-hexyl-2-{3-[(2Z)-3-hexyl-2,3-dihydro-1,3-benzoxazol-2-ylidene]prop-1-en-1-yl}-1,3-benzoxazol-3-ium iodide
Synonyms: DiOC6
Molecular Weight (g/mol): 572.53
Molecular Formula: C29H37IN2O2
InChi Key: XVLXYDXJEKLXHN-UHFFFAOYSA-M
SMILES: [I-].CCCCCCN1\C(OC2=CC=CC=C12)=C\C=CC1=[N+](CCCCCC)C2=CC=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results