Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Dihexyloxacarbocyanine Iodide 98%, Thermo Scientific™

Catalog No. AC407600010
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC407600010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC407600010 Supplier Thermo Scientific Chemicals Supplier No. 407600010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 53213-82-4
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C29H37IN2O2
Quantity 1g
MDL Number MFCD00011954
Synonym 3,3'-dihexyloxacarbocyanine iodide
InChI Key XVLXYDXJEKLXHN-UHFFFAOYSA-M
SMILES [I-].CCCCCCN1\C(OC2=CC=CC=C12)=C\C=CC1=[N+](CCCCCC)C2=CC=CC=C2O1
IUPAC Name 3-hexyl-2-{3-[(2Z)-3-hexyl-2,3-dihydro-1,3-benzoxazol-2-ylidene]prop-1-en-1-yl}-1,3-benzoxazol-3-ium iodide
Molecular Weight (g/mol) 572.53
PubChem CID 129893825
Formula Weight 572.52
Percent Purity ≥97.5%
Chemical Name or Material 3, 3'-Dihexyloxacarbocyanine iodide, 98%
Show More Show Less
Hazard Category LIGHT SENSITIVE
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.