accessibility menu, dialog, popup

UserName

Viewing profile as user:
IH3CNSSNH3C(Z)(E)(E/Z)

CAS RN 53213-94-8

IUPAC Name: 3-propyl-2-[(3E)-5-[(2Z)-3-propyl-2,3-dihydro-1,3-benzothiazol-2-ylidene]penta-1,3-dien-1-yl]-1,3-benzothiazol-3-ium iodide
Synonyms: 3-propyl-2-[(3E)-5-[(2Z)-3-propyl-1,3-benzothiazol-2-ylidene]penta-1,3-dien-1-yl]-1,3-benzothiazol-3-ium iodide
Molecular Weight (g/mol): 546.53
Molecular Formula: C25H27IN2S2
InChi Key: GDEURKKLNUGTDA-UHFFFAOYSA-M
SMILES: [I-].CCCN1\C(SC2=CC=CC=C12)=C\C=C\C=CC1=[N+](CCC)C2=CC=CC=C2S1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)