Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Di-n-propylthiadicarbocyanine iodide, 96%

Catalog No. AAH34209MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34209MD 250 mg
AAH3420903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH34209MD Supplier Thermo Scientific Chemicals Supplier No. H34209MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 53213-94-8 | C25H27IN2S2 | 546.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53213-94-8
Molecular Formula C25H27IN2S2
Molecular Weight (g/mol) 546.53
MDL Number MFCD00075033,MFCD00075033
InChI Key GDEURKKLNUGTDA-UHFFFAOYSA-M
Synonym disc3 5, 3,3'-dipropylthiadicarbocyanine iodide, 3-propyl-2-5-3-propyl-2 3h-benzothiazolylidene-1,3-pentadienyl benzothiazolium iodide, 5-3-propylbenzothiazol-2-ylidene-1-3-propylbenzothiazolium-2-yl-1,3-pentadiene iodide
PubChem CID 91972132
SMILES [I-].CCCN1\C(SC2=CC=CC=C12)=C\C=C\C=CC1=[N+](CCC)C2=CC=CC=C2S1

Specifications

Melting Point >250°C
Quantity 250 mg
Sensitivity Light sensitive
IUPAC Name (2Z)-3-propyl-2-[5-(3-propyl-1,3-benzothiazol-3-ium-2-yl)penta-2,4-dienylidene]-1,3-benzothiazole;iodide
Formula Weight 546.54
Percent Purity 96%
Chemical Name or Material 3,3'-Di-n-propylthiadicarbocyanine iodide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.