accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 532391-31-4

IUPAC Name: 2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 235.09
Molecular Formula: C12H18BNO3
InChi Key: JEJYNUCGRAHMDO-UHFFFAOYSA-N
SMILES: COC1=NC=CC=C1B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results