accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5325-64-4

IUPAC Name: hydrogen 2-(methylamino)acetamide chloride
Synonyms: hydrogen 2-(methylamino)acetamide chloride
Molecular Weight (g/mol): 124.57
Molecular Formula: C3H9ClN2O
InChi Key: VVIXOTCTYAILNP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CNCC(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results