accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNHNNNO(R)OH(S)HO(R)HO

CAS RN 53296-10-9

IUPAC Name: (3R,4S,5R)-2-[6-amino-2-(phenylamino)-9H-purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: (3R,4S,5R)-2-[6-amino-2-(phenylamino)purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol): 358.36
Molecular Formula: C16H18N6O4
InChi Key: SCNILGOVBBRMBK-FJFSNTMWSA-N
SMILES: NC1=C2N=CN(C3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC(NC2=CC=CC=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results