CAS RN 533-50-6
IUPAC Name:
(3S)-1,3,4-trihydroxybutan-2-one
Synonyms:
L-erythrulose
Molecular Weight (g/mol):
120.1
Molecular Formula:
C4H8O4
InChi Key:
UQPHVQVXLPRNCX-YEQUQEJONA-N
SMILES:
OC[C@H](O)C(=O)CO
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals L-Erythrulose
Thermo Scientific Chemicals L-Erythrulose
CAS: 533-50-6 Molecular Formula: C4H8O4