accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSH

CAS RN 5332-52-5

IUPAC Name: undecane-1-thiol
Synonyms: 1-undecanethiol
Molecular Weight (g/mol): 188.37
Molecular Formula: C11H24S
InChi Key: CCIDWXHLGNEQSL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCS
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)