Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Undecanethiol

Catalog No. 5104675G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-046-75G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 51-046-75G Supplier Sigma Aldrich Supplier No. 5104675G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Undecanethiol (UDT) is an alkanethiol that forms a self-assembled monolayer (SAM) on a variety of surfaces. It immobilizes the surface atoms and enhances the absorption of proteins. It can be synthesized by reacting bromo-1-undecanol and thiourea and subsequent purification using ethyl acetate.UDT can be used to functionalize polycarbonate filter membrane for immunological experiments which can further be used in the fabrication of plasmonic flow through biosensor based applications. SAM of UDT can be used in the colloidal gold enhanced immunoassay. It may be used to form SAM on quartz crystal microbalance (QCM) gold electrodes which can be used as adhesive intermediate layers.

Specifications

CAS 5332-52-5
Density 0.841 g/mL (at 25°C)
Boiling Point 103°C to 104°C (3 mmHg)
Molecular Formula C11H24S
Refractive Index n20/D 1.459 (literature)
Linear Formula CH3(CH2)10 SH
MDL Number MFCD00022097
Quantity 5 g
Synonym 1-UDT; 11-Mercapto-1-undecanol; Undecyl Mercaptan
Molecular Weight (g/mol) 188.37
Percent Purity 98%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.